siRNA Details
| Gene Symbol: LOC100996842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322800.1 | NP_001309729.1 | 2 | 1039 | ||
| NR_136401.1 | 2 | 582 | ||||
| NR_136402.1 | 2 | 214 | ||||
| NR_136403.1 | 2 | 671 | ||||
| GenBank mRNA | BM888151.1 | - | 2 | 203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417897.1 | - | 2 | 203 | Chr. 17 - 7582016 - 7584068 [-] on Build GRCh38 | |

LOC100996842
Genomic Map