siRNA Details
| Gene Symbol: LOC101929464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125978.1 | 2 | 168 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432703.1 | - | 2 | 168 | Chr. 1 - 33350352 - 33363245 [+] on Build GRCh38 | |

LOC101929464
Genomic Map