siRNA Details
| Gene Symbol: LOC100129434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125368.1 | 3 | 847 | |||
| GenBank mRNA | AK096196.1 | - | 3 | 1059 | ||
| AK311113.1 | - | 3 | 847 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129434| Gene Symbol: LOC100129434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125368.1 | 3 | 847 | |||
| GenBank mRNA | AK096196.1 | - | 3 | 1059 | ||
| AK311113.1 | - | 3 | 847 | |||