siRNA Details
Gene Symbol: LOC105372626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134577.1 | 2 | 281 | |||
GenBank mRNA | BX089899.1 | - | 2 | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000428529.1 | - | 2 | 279 | Chr. 20 - 43894720 - 43895468 [+] on Build GRCh38 |
