siRNA Details
| Gene Symbol: LOC101928401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108099.1 | 2 | 2613 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429367.1 | - | 2 | 2613 | Chr. 7 - 56534230 - 56538130 [-] on Build GRCh38 | |

LOC101928401
Genomic Map