siRNA Details
Gene Symbol: LINC01346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040065.1 | 3 | 468 | |||
GenBank mRNA | AK124708.1 | - | 3 | 463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412674.1 | - | 3 | 463 | Chr. 1 - 3940617 - 3955262 [+] on Build GRCh38 |
