siRNA Details
| Gene Symbol: LINC01346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124708.1 | - | 3 | 2048 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412674.1 | - | 3 | 2048 | Chr. 1 - 3940617 - 3955262 [+] on Build GRCh38 | |

LINC01346
Genomic Map