siRNA Details
| Gene Symbol: LINC00028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024358.1 | 4 | 568 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435497.1 | - | 4 | 568 | Chr. 20 - 31485778 - 31487574 [+] on Build GRCh38 | |

LINC00028
Genomic Map