siRNA Details
| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093142.1 | - | 1 | 416 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451929.1 | - | 1 | 417 | Chr. 10 - 43939003 - 43944466 [+] on Build GRCh38 | |

LINC00841
Genomic Map