siRNA Details
| Gene Symbol: LOC100506077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX119135.1 | - | 1 | 170 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432251.1 | - | 1 | 172 | Chr. 2 - 162093429 - 162094854 [+] on Build GRCh38 | |

LOC100506077
Genomic Map