siRNA Details
Gene Symbol: LOC100506077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX119135.1 | - | 2 | 286 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000432251.1 | - | 2 | 288 | Chr. 2 - 162093429 - 162094854 [+] on Build GRCh38 |
