siRNA Details
| Gene Symbol: LOC100128317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126025.1 | 11 | 1804 | |||
| NR_126026.1 | 9 | 1517 | ||||
| GenBank mRNA | AY927633.1 | - | 11 | 1803 | ||
| AY927634.1 | - | 9 | 1517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128317| Gene Symbol: LOC100128317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126025.1 | 11 | 1804 | |||
| NR_126026.1 | 9 | 1517 | ||||
| GenBank mRNA | AY927633.1 | - | 11 | 1803 | ||
| AY927634.1 | - | 9 | 1517 | |||