siRNA Details
| Gene Symbol: LOC100128317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126025.1 | 6 | 641 | |||
| NR_126026.1 | 5 | 595 | ||||
| GenBank mRNA | AY927633.1 | - | 6 | 641 | ||
| AY927634.1 | - | 5 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128317| Gene Symbol: LOC100128317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126025.1 | 6 | 641 | |||
| NR_126026.1 | 5 | 595 | ||||
| GenBank mRNA | AY927633.1 | - | 6 | 641 | ||
| AY927634.1 | - | 5 | 595 | |||