siRNA Details
| Gene Symbol: ZNF529-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110703.1 | 1, 2 | 464 | |||
| NR_110705.1 | 1, 2 | 464 | ||||
| NR_110706.1 | 1, 2 | 464 | ||||
| GenBank mRNA | BC039524.1 | - | 1, 2 | 464 | ||
| CD248632.1 | - | 1, 2 | 142 | |||
| CK820071.1 | - | 1, 2 | 157 | |||

ZNF529-AS1
Genomic Map