siRNA Details
Gene Symbol: ZNF529-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110705.1 | 4 | 681 | |||
GenBank mRNA | BC039524.1 | - | 4 | 681 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000475219.6 | - | 4 | 681 | Chr. 19 - 36573070 - 36594708 [+] on Build GRCh38 |
