siRNA Details
| Gene Symbol: ZNF529-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110703.1 | 4 | 855 | |||
| NR_110704.1 | 4 | 885 | ||||
| NR_110705.1 | 5 | 999 | ||||
| GenBank mRNA | BC039524.1 | - | 5 | 999 | ||
| CD248632.1 | - | 4 | 533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF529-AS1| Gene Symbol: ZNF529-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110703.1 | 4 | 855 | |||
| NR_110704.1 | 4 | 885 | ||||
| NR_110705.1 | 5 | 999 | ||||
| GenBank mRNA | BC039524.1 | - | 5 | 999 | ||
| CD248632.1 | - | 4 | 533 | |||