siRNA Details
| Gene Symbol: LOC730338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134575.1 | 3 | 988 | |||
| GenBank mRNA | AK125311.1 | - | 3 | 988 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451905.4 | - | 3 | 988 | Chr. 7 - 46687879 - 46697122 [-] on Build GRCh38 | |

LOC730338
Genomic Map