siRNA Details
Gene Symbol: LINC00690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103843.1 | 2 | 290 | |||
GenBank mRNA | BC034913.1 | - | 2 | 293 | ||
BX100558.1 | - | 2 | 290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429949.1 | - | 2 | 288 | Chr. 3 - 16536320 - 16541373 [+] on Build GRCh38 |
