siRNA Details
| Gene Symbol: LOC102723833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125992.1 | 3 | 337 | |||
| NR_125993.1 | 2 | 208 | ||||
| GenBank mRNA | BX340080.2 | - | 3 | 337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723833| Gene Symbol: LOC102723833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125992.1 | 3 | 337 | |||
| NR_125993.1 | 2 | 208 | ||||
| GenBank mRNA | BX340080.2 | - | 3 | 337 | ||