siRNA Details
Gene Symbol: LOC102723833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125992.1 | 1, 2 | 145 | |||
GenBank mRNA | BX340080.2 | - | 1, 2 | 145 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446411.5 | - | 1, 2 | 145 | Chr. 1 - 216072465 - 216086917 [+] on Build GRCh38 |
