siRNA Details
Gene Symbol: LOC105374928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030116.1 | - | 3 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419796.1 | - | 3 | 653 | Chr. 6 - 11417350 - 11481239 [+] on Build GRCh38 |
