siRNA Details
| Gene Symbol: LOC101929122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121668.1 | 4 | 664 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419064.3 | - | 2 | 238 | Chr. 6 - 159165909 - 159167353 [-] on Build GRCh38 | |

LOC101929122
Genomic Map