siRNA Details
| Gene Symbol: LINC01122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033873.1 | 6, 7 | 834 | |||
| GenBank mRNA | AK055400.1 | - | 6, 7 | 834 | ||
| AL534733.3 | - | 3, 4 | 567 | |||
| AL832565.1 | - | 1, 2 | 38 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427421.5 | - | 6, 7 | 834 | Chr. 2 - 58520753 - 59061653 [+] on Build GRCh38 | |

LINC01122
Genomic Map