siRNA Details
| Gene Symbol: LINC00284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001938.1 | - | 1 | 21 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000423211.1 | - | 1 | 21 | Chr. 13 - 44025319 - 44029937 [+] on Build GRCh38 | |

LINC00284
Genomic Map