siRNA Details
Gene Symbol: LINC00284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026955.1 | 1 | 54 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000592085.1 | - | 1 | 54 | Chr. 13 - 44022335 - 44028548 [+] on Build GRCh38 |
