siRNA Details
| Gene Symbol: LINC00656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034149.1 | 4 | 1410 | |||
| GenBank mRNA | AF258576.1 | - | 4 | 1397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443744.1 | - | 4 | 1395 | Chr. 20 - 23125068 - 23132621 [-] on Build GRCh38 | |

LINC00656
Genomic Map