siRNA Details
| Gene Symbol: LOC100506289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537871.1 | - | 2 | 1430 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420912.1 | - | 2 | 1430 | Chr. 7 - 26551822 - 26557200 [+] on Build GRCh38 | |

LOC100506289
Genomic Map