siRNA Details
| Gene Symbol: LINC01251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109755.1 | 2 | 221 | |||
| GenBank mRNA | BX115224.1 | - | 2 | 228 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444956.5 | - | 2 | 221 | Chr. 9 - 33732975 - 33738416 [-] on Build GRCh38 | |

LINC01251
Genomic Map