siRNA Details
| Gene Symbol: LOC100506098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134567.1 | 5 | 726 | |||
| NR_134568.1 | 5 | 635 | ||||
| NR_134569.1 | 4 | 564 | ||||
| GenBank mRNA | BI759100.1 | - | 5 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506098| Gene Symbol: LOC100506098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134567.1 | 5 | 726 | |||
| NR_134568.1 | 5 | 635 | ||||
| NR_134569.1 | 4 | 564 | ||||
| GenBank mRNA | BI759100.1 | - | 5 | 626 | ||