siRNA Details
Gene Symbol: KCNQ5-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120503.1 | 2 | 137 | |||
GenBank mRNA | BX104605.1 | - | 2 | 137 | ||
HY037383.1 | - | 2 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445310.1 | - | 2 | 137 | Chr. 6 - 72630495 - 72678558 [+] on Build GRCh38 |
