siRNA Details
| Gene Symbol: RIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005260731.2 | XP_005260788.1 | 2 | 367 | ||
| XM_006723574.3 | XP_006723637.1 | 2 | 135 | |||
| XM_006723575.3 | XP_006723638.1 | 2 | 135 | |||
| XM_017027887.1 | XP_016883376.1 | 2 | 332 | |||
| XM_017027888.1 | XP_016883377.1 | 2 | 332 | |||
| GenBank mRNA | BC090059.1 | - | 1 | 85 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412571.1 | - | 2 | 217 | Chr. 20 - 19758148 - 19809282 [+] on Build GRCh38 | |

RIN2
Genomic Map