siRNA Details
| Gene Symbol: KRTAP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021489.1 | 2 | 756 | |||
| GenBank mRNA | AK055141.1 | - | 2 | 447 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424148.1 | - | 2 | 755 | Chr. 11 - 1572741 - 1599184 [+] on Build GRCh38 | |

KRTAP5-AS1
Genomic Map