siRNA Details
| Gene Symbol: LOC101929526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109955.1 | 4 | 591 | |||
| GenBank mRNA | BC042559.1 | - | 4 | 591 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428285.1 | - | 4 | 592 | Chr. 20 - 18674395 - 18698709 [-] on Build GRCh38 | |

LOC101929526
Genomic Map