siRNA Details
| Gene Symbol: LINC01425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109958.1 | 5 | 589 | |||
| GenBank mRNA | BC035642.1 | - | 5 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01425| Gene Symbol: LINC01425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109958.1 | 5 | 589 | |||
| GenBank mRNA | BC035642.1 | - | 5 | 599 | ||