siRNA Details
Gene Symbol: LOC101928403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135540.1 | 1 | 603 | |||
GenBank mRNA | BC017749.1 | - | 1 | 261 | ||
BC046476.1 | - | 1 | 603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000453008.2 | - | 1 | 893 | Chr. 2 - 88627539 - 88631819 [+] on Build GRCh38 |
