siRNA Details
| Gene Symbol: LINC01505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104145.1 | 5 | 894 | |||
| GenBank mRNA | AK093363.1 | - | 5 | 894 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444985.6 | - | 5 | 894 | Chr. 9 - 106616058 - 106679798 [+] on Build GRCh38 | |

LINC01505
Genomic Map