siRNA Details
Gene Symbol: LINC01505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104145.1 | 5 | 1471 | |||
GenBank mRNA | AK093363.1 | - | 5 | 1471 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444985.6 | - | 5 | 1471 | Chr. 9 - 106616058 - 106679798 [+] on Build GRCh38 |
