siRNA Details
| Gene Symbol: DEPDC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110671.1 | 3 | 1131 | |||
| GenBank mRNA | BC016787.1 | - | 3 | 1135 | ||
| BC020917.1 | - | 3 | 1111 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428732.1 | - | 3 | 1131 | Chr. 1 - 68496676 - 68538627 [+] on Build GRCh38 | |

DEPDC1-AS1
Genomic Map