siRNA Details
| Gene Symbol: DEPDC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110671.1 | 1, 2 | 104 | |||
| GenBank mRNA | BC016787.1 | - | 1, 2 | 108 | ||
| BC020917.1 | - | 1, 2 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEPDC1-AS1| Gene Symbol: DEPDC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110671.1 | 1, 2 | 104 | |||
| GenBank mRNA | BC016787.1 | - | 1, 2 | 108 | ||
| BC020917.1 | - | 1, 2 | 84 | |||