siRNA Details
| Gene Symbol: LOC100505515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109870.1 | 1 | 168 | |||
| GenBank mRNA | HY040996.1 | - | 1 | 169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505515| Gene Symbol: LOC100505515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109870.1 | 1 | 168 | |||
| GenBank mRNA | HY040996.1 | - | 1 | 169 | ||