siRNA Details
| Gene Symbol: LINC00229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_044991.1 | 2 | 203 | |||
| GenBank mRNA | BX090786.1 | - | 2 | 203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443783.1 | - | 2 | 203 | Chr. 22 - 44606939 - 44625419 [-] on Build GRCh38 | |

LINC00229
Genomic Map