siRNA Details
| Gene Symbol: LOC101929723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110262.1 | 1 | 31 | |||
| NR_110263.1 | 1 | 31 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436551.1 | - | 1 | 31 | Chr. 2 - 41931619 - 41933723 [-] on Build GRCh38 | |

LOC101929723
Genomic Map