siRNA Details
Gene Symbol: LINC01049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120414.1 | 2 | 143 | |||
GenBank mRNA | BC038529.1 | - | 2 | 143 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426840.1 | - | 2 | 144 | Chr. 13 - 90493287 - 90535080 [+] on Build GRCh38 |
