siRNA Details
| Gene Symbol: LINC01250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110228.1 | 7 | 2190 | |||
| GenBank mRNA | AK095310.1 | - | 7 | 2190 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457478.1 | - | 7 | 2190 | Chr. 2 - 2895048 - 3126026 [-] on Build GRCh38 | |

LINC01250
Genomic Map