siRNA Details
Gene Symbol: LINC01250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110228.1 | 7 | 1117 | |||
GenBank mRNA | AK095310.1 | - | 7 | 1117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457478.1 | - | 7 | 1117 | Chr. 2 - 2895048 - 3126026 [-] on Build GRCh38 |
