siRNA Details
Gene Symbol: MIR3663HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121650.1 | 2 | 1432 | |||
GenBank mRNA | BC039338.1 | - | 2 | 1432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419373.2 | - | 2 | 1436 | Chr. 10 - 117158656 - 117169059 [-] on Build GRCh38 |
