siRNA Details
Gene Symbol: ERICH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121670.1 | 3 | 467 | |||
NR_121671.1 | 3 | 374 | ||||
GenBank mRNA | BX092187.1 | - | 3 | 467 | ||
CR627203.1 | - | 3 | 374 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ERICH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121670.1 | 3 | 467 | |||
NR_121671.1 | 3 | 374 | ||||
GenBank mRNA | BX092187.1 | - | 3 | 467 | ||
CR627203.1 | - | 3 | 374 |