siRNA Details
Gene Symbol: LINC01506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109769.1 | 3, 4 | 292 | |||
GenBank mRNA | CR745134.1 | - | 3, 4 | 292 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109769.1 | 3, 4 | 292 | |||
GenBank mRNA | CR745134.1 | - | 3, 4 | 292 |