siRNA Details
| Gene Symbol: LOC102724357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134619.1 | 1, 2 | 196 | |||
| GenBank mRNA | BX096530.1 | - | 1, 2 | 196 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413324.1 | - | 1, 2 | 164 | Chr. 6 - 169067818 - 169069416 [+] on Build GRCh38 | |

LOC102724357
Genomic Map