siRNA Details
| Gene Symbol: SYNE1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120501.1 | 2 | 373 | |||
| GenBank mRNA | CK821189.1 | - | 2 | 373 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412161.1 | - | 2 | 357 | Chr. 6 - 152380546 - 152381564 [+] on Build GRCh38 | |

SYNE1-AS1
Genomic Map